missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pre-cut Blank Labels for BBP3X, S3XXX, and i3300 Printers
The material of these labels is ideal for indoor and outdoor facility labels.
£118.00 - £265.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Finish | Gloss |
Material | Vinyl (B-7569) |
Print Technology | Thermal Transfer |
Product Code | Brand | Color | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
15991001
|
Brady™
120932 |
White | 72 mm | 82.6 mm | 28.6 mm |
£118.00
Pack of 300 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15981001
|
Brady™
120931 |
White | 75 mm | 82.6 mm | 57 mm |
£171.00
Pack of 300 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15911011
|
Brady™
120935 |
Yellow | 75 mm | 82.6 mm | 57 mm |
£171.00
Pack of 300 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15971001
|
Brady™
120930 |
White | 152 mm | 158.8 mm | 101 mm |
£234.72
Pack of 175 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15901011
|
Brady™
120934 |
Yellow | 152 mm | 158.8 mm | 101 mm |
£265.00
Pack of 175 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
Description
- Ideal for 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
- Vinyl material
Specifications
Not Autoclavable | |
Gloss | |
Thermal Transfer | |
Facility Identification, Hazard Communication Identification, Electrical Labeling | |
1 | |
-20°C to +75°C |
Permanent Acrylic | |
Vinyl (B-7569) | |
Non-sterile | |
BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, S3000, S3100, and i5300 Industrial Label Printers | |
Rectangle | |
0.102 mm |