missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precision™ Shaking Water Baths

Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.

Brand:  Thermo Scientific™ TSSWB15S

 View more versions of this product

Product Code. 15385867

  • £2178.31 / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
TRUSTED_SUSTAINABILITY
Specifications

Specifications

304 Stainless Steel
15L Shallow Shaking Water Bath
UL, CE
Monochrome LCD
Stainless Steel Gable Cover and Rubber Duck
Precision SWB 15S
±0.1°C
15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
9.0-10.5 / 4.3-5.0 (120V/230V)
Digital
15 L
Ambient +5° to 100°C
1200 w
Epoxy Powder-coated Cold Rolled Steel
4
±0.05°C
Global Voltage 100/115 V 200/230 V 50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers