Learn More
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
Brand: Ark-Plas Products™ MAD60-DF0
This item is not returnable.
View return policy
Description
- Made of Kynar
- Economical straight barbed
- Easily connect fluid paths without the use of tools
Specifications
Straight reducing connector | |
Fluororesin | |
3/16 x 1/16 in. | |
Natural |