Learn More
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
£17.00
Spécification
Product Type | Straight reducing connector |
---|---|
Material | Fluororesin |
Color | Natural |
Code produit | Marque | Tubing Size | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Tubing Size | Prix | Quantité et disponibilité | |||||
12313850
|
Ark-Plas Products™
MAD80-DF0 |
5/32 x 1/16 in. |
£17.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
12323850
|
Ark-Plas Products™
MAD60-DF0 |
3/16 x 1/16 in. |
£17.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Made of Kynar
- Economical straight barbed
- Easily connect fluid paths without the use of tools
Spécification
Straight reducing connector | |
Fluororesin | |
Natural |