missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PreScission™ Protease

Use for site-specific separation of the GST tag from proteins expressed using pGEX-6P vectors. Cytiva Life Sciences™ PreScission™ Protease G allows simultaneous protease immobilization and cleavage of GST fusion proteins.

Brand:  Cytiva 27-0843-01

Product Code. 10196324

  • £298.00 / 500 units
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Description: A genetically engineered fusion protein consisting of human rhinovirus 3C protease and GST; Specifically cleaves between the Gln and Gly residues of the recognition sequence of LeuGluValLeuPheGln/GlyPro.
  • Application: Facilitates removal of the protease by allowing simultaneous protease immobilization and cleavage of GST fusion proteins produced from the pGEX-6P vectors pGEX-6P-1, pGEX-6P-2, and pGEX-6P-3
  • Storage Buffer Components: 50mM Tris-HCl (pH 8.0), 150mM NaCl, 10mM EDTA, 1mM DTT, and 20% glycerol.
  • Unit Definition: One unit will cleave >90% of 100μg of a test GST-fusion protein in 50mM Tris-HCl, 150mM NaCl, 1mM EDTA, 1mM DTT, pH 7.0 at 5°C for 16 hrs.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Elution Buffer: 50mM Tris HCl, 10mM reduced Glutathione pH 8; Cleavage Buffer: 50mM Tris HCl pH 8, 150mM NaCl, 10mM EDTA, 1mM DTT and 20% Glycerol
500U
Facilitates removal of the protease by allowing simultaneous protease immobilization and cleavage of >90% of 100μg of a test GST-fusion protein produced from the pGEX-6P vectors pGEX-6P-1, pGEX-6P-2 and pGEX-6P-3
Fusion Protein Protease
-20°C/-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers