missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Cell Lysis Buffer

Used to lyse cells under nondenaturing conditions. Invitrogen™ ProcartaPlex™ Cell Lysis Buffer is available in 20mL units that should be stored chilled (for continual use) or frozen (for longer periods of time.) Additional protease inhibitors can be added without any difficulties.

Brand:  Invitrogen™ EPX-99999-000

Product Code. 15572117

  • £153.00 / 20mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For lysis of adherent cells, 400μL of cell lysis buffer is used for a 10cm dish. For non-adherent cells, 400μL of buffer is used per 107 cells, once they have been washed in PBS and pelleted. Buffer should be chilled and PMSF should be added to the buffer immediately prior to use.

Buffer composition:

  • 20mM Tris-HCl (pH 7.5)
  • 150mM NaCl
  • 1mM Na2EDTA
  • 1mM EGTA
  • 1% Triton
  • 2.5mM sodium pyrophosphate
  • 1mM β-glycerophosphate
  • 1mM Na3VO4
  • 1μg/ml leupeptin
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer
Canine, Mouse, Rat, Porcine, Non-Human Primate, Human
ProcartaPlex™
20 mL
Luminex™ Instruments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number442419-000 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog Number
Lot Number429592-000 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberEPX-99999-000
Lot Number422413-000 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog NumberEPX-99999-000
Lot Number417331-000 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog NumberEPX-99999-000
Lot Number413129-000 Certificate TypeCertificate of Analysis Date10/06/2024 Catalog NumberEPX-99999-000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ProcartaPlex™ Cell Lysis Buffer > ProcartaPlex Cell Lysis Buffer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.