Learn More
Invitrogen™ ProcartaPlex™ Wash Buffer (10X)
ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.
Brand: Invitrogen™ EPX-66666-000
Description
ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.
ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.
Reported application
- Multiplex immunoassay

Specifications
Buffer | |
25 mL | |
Luminex™ Instruments |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
442219-000 | Certificate of Analysis | 10/04/2025 | |
438176-000 | Certificate of Analysis | 20/03/2025 | |
438176-000 | Certificate of Analysis | 10/02/2025 | |
438176-000 | Certificate of Analysis | 25/01/2025 | EPX-66666-000 |
425499-000 | Certificate of Analysis | 02/10/2024 | EPX-66666-000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.