missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Wash Buffer (10X)

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.

Brand:  Invitrogen™ EPX-66666-001

 View more versions of this product

Product Code. 16396622

  • £277.00 / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.

ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.

Reported application

  • Multiplex immunoassay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer
200 mL
Luminex™ Instruments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number440771-000 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number439702-000 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number
Lot Number440771-000 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog Number
Lot Number439702-000 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog Number
Lot Number440771-000 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ProcartaPlex™ Wash Buffer (10X) > 200 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.