Learn More
Elma™ Modular Baskets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
Brand:  Elma™ 3000006012
 View more versions of this product
Product Code. 17821337
  • £18.30 / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Small Basket
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
16 mm
64 mm
1 Section
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Elma™ Modular Baskets > Modular Baskets and Lids; Diameter (Metric): 64 mm; Height (Metric): 16 mm; Section: 1 Section