Learn More
Elma Basket System
Modular basket system for Elmasonic P, Elmasonic Select, EASY
Brand:  Elma 1108916
 View more versions of this product
Product Code. 17891666
  • £47.55 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Modular Basket System
2.2 lb.
105 x 195 x 50 mm
1.0 kg
Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Basket System Modular Type 30
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Elma Basket System > Model: Basket System Modular type 10