Learn More
BD Biosciences Reagent Rack - APC
Pre-cooled reagent rack keeps your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability.
Brand:  BD Biosciences 657181
Product Code. 15921270
  • £59.40 / Each
Explore more special offers
Description
Description
Pre-cooled BD™ Reagent Racks keep your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability. The BD Reagent Racks are stackable and available in different colors to help you keep your vials organized on the bench as well as in the fridge. Each rack is filled with cooling material, and can accommodate up to 25 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials. Size 180 mm x 54,5 mm - Weight: 950 gr.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Filled with cooling material.
Yes
950 G
BD™ Amber Vials (5) and Narrower BD Pharmingen™ Type Vials (20)
180 mm x 54.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Biosciences Reagent Rack - APC > Holds: 5 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials.