Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand: Fisherbrand™ 1003281
Description
- Easy-to-open and close cover
- Muffles noise level
- Designed to be used as drip basin when flipped over
- Chemical-resistant plastic
- Bring your ultrasonic cleaner up to temperature more quickly
- Contain and prevent spills and leakage
Specifications
Lid | |
Ultrasonic Cleaner, 2.75 and 4.25 L | |
Medical Laboratories and in Industrial Production Processes | |
Plastic |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
Product Content Correction
Product Title
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners > For use with FB11201 Ultrasonic Unit
Spot an opportunity for improvement?Share a Content Correction