Learn More
Hoefer™ HE 33 Mini Submarine Electrophoresis Unit
Designed for very fast separations of DNA restriction fragments in agarose gels
Brand:  Hoefer™ HE33B
Code : NEW
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 10646253
  • £368.28 / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Includes: Buffer chamber, lid with cables, UV-transparent gel running tray, gel casting tray, 4 foam gaskets, bubble level. Complete system (03-500-130) also includes 1.5mm thick 8-well comb and comb back.
Description
Description
Designed for very fast separations of DNA restriction fragments in agarose gels
  • The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
  • Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
  • No taping required for casting
  • Electrical safety locks in lid
Specifications
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Up to 80%
Up to 32
50°C
250 mL
9.4 in.
24 cm
5.1 in.
13 cm
2.8 in.
500 V
500 mA
Mini Horizontal Agarose Electrophoresis Unit
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level
122°F
8.4 oz.
9.4 x 5.1 x 2.8 in. (24 x 13 x 7 cm)
3.9 in.
10 cm
2.7 in.
7 cm
7 cm
15 w