missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets

Conserve energy with these cabinets, which combine smart design and extraordinary value with best-in-class energy efficiency, reliability and usability.

Brand:  Thermo Scientific™ 51028225


6703.43 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : 50

Additional Details : Weight : 184.50000kg

 View more versions of this product

Product Code. 11762997

  • £8805.37 / Each
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets (BSCs) combine safety and extraordinary value with energy efficiency, reliability and usability. With proprietary airflow design and energy-efficient DC motors, the MSC-Advantage BSCs use up to 68% less energy than cabinets with traditional AC motors. The cabinets provide both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. Cabinets are certified to EN 12469 safety standard.

Features:

  • Available in 90cm, 120cm, 150cm and 180cm
  • With microprocessor control
  • Large front-panel display (easily visible from seated position) provides constant readout of downflow and inflow velocities and filter usage status
  • Exclusive Performance Factor monitoring system advises when cabinet service is needed
  • Proprietary window opens to 20cm working height, 53.5cm maximum height, easily lowers for interior cleaning
  • Ergonomically angled window is sloped at 10° for better comfort and reduced operator fatigue
  • Unique airflow technology and intelligent motor design dramatically improve energy efficiency, operation and serviceability; provide excellent sample protection
  • SmartPort: T¨UV-approved, safe, clean method for vacuum tubing and cables routing method through the side wall of the biological safety cabinet
  • Supply/exhaust air filter: HEPA H 14 EN 1822, 99.995% at 0.3μm particle size

Construction:

  • Front-accessible components allow for quick change-outs of HEPA filters and other components Quiet operation at <55dBA enables a comfortable working environment
  • Divided stainless-steel work tray removes easily for cleaning or autoclaving
  • Standard armrests designed to prevent accidental blocking with coat sleeves
  • Very large work surface (0.56m2 or 0.84m2) improves productivity
  • Smooth components throughout the cabinet virtually eliminate risk of injury during routine cleaning, servicing or maintenance

Certifications/Compliance

  • Fully compliant with the EN 12469 safety standard as independently tested and certified by TUV Nord

Ordering Information:

Accessories include programmable UV light, one-piece work tray, stand options, Ergolign saddle stool, free-standing floor shelf. Not available for sale in North America.

Specifications

Specifications

EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m)
Biosafety Cabinet
1 Double, Right Side
HEPA H 14 EN 1822, 99.995% at 0.3μm Particle
Energy efficient
TUV-approved
152 cm
230 V
0.9 m
<55 dBA
39.3 in.
100 cm
<40 W
2.95 ft.
80 cm
150 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services