missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (10 mg/mL)

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

Brand:  Gibco™ 15710064


 View more versions of this product

Product Code. 15820243

  • £14.86 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria.

  • Effective against wide variety of gram-positive and gram-negative bacteria
  • Used for prevention of bacterial contamination on cell cultures
  • Recommended working concentration ranges from 0.5 to 50μg/mL

Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Mammalian Cell Culture
Antibiotic
Prevention of Cell Culture Contamination
Room Temperature
10 mg/mL
Liquid
Sterile-filtered
10 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gentamicin Reagent Solution

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : gentamicin sulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.