missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

Brand:  Thermo Scientific™ SPD300DDA-230


23836.43 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 73.00000kg

 View more versions of this product

Product Code. 15936181

  • £29428.67 / Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket

Description

Description

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

  • For strong acids and bases used in combinatorial chemistry
Specifications

Specifications

UL Listed to US and Canadian requirements. CE marked
Aluminum
35°C to 85°C
SpeedVac SPD300DDA Vacuum concentrator
SPD300
230 V 50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services