Learn More
Sartorius Cubis™ II Precision (1-place) Balance, MCE Interface
Cubis™ II Precision (1-Place) Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCE12201S0-S00
Additional Details : Weight : 6.77000kg
 View more versions of this product
Product Code. 16223252
  • £5050.00 / 1 unit
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report, QP99 Software
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±0.05 g
Precision (1-place) Balance
425 mm
95 mm
120/240 V
50/60 Hz (± 5%)
12,200 g
20 mg
±30 mg
No draft shield
0.8 s
10 years
240 mm
5.4 kg
100/240 V