Learn More
Sartorius Cubis™ II High-Capacity Lab Balance
The Cubis™ II High-Capacity Lab Balance with a maximum load between 11.2 kg and 70.2 kg and readability between 0.1 g and 1 g provides the ideal model for every application.
Brand:  Sartorius MCA70200S-2S00-0 QP1
Additional Details : Weight : 17.10000kg
 View more versions of this product
Product Code. 16497025
  • £10770.00 / Each
Estimated Shipment: 11-09-2024
to see stock.
Description
Description
The New Generation of Modular Premium Laboratory Balances
  • Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
  • One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
  • All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
  • These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
  • It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Specifications
Balance with Type Approval Certificate for China
70,200 g
100 mg
Aluminum, plastic PBT, Optiwhite float glass and stainless steel
1.5 sec.
100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A
D-sub connector, 9-pin
Color Touchscreen
Draft Shield O, QP1
S00 Standard version non-verified
Electronic Toploading Balance