missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader

The BioTek Synergy LX multi-mode reader automates many common microplate assays. For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek SLXF-SN


12625.50 GBP valid until 2024-08-31
Use promo code "21622" to get your promotional price.



Additional Details : Weight : 19.00000kg

 View more versions of this product

Product Code. 17287829

  • £13290.00 / Each
Estimated Shipment: 14-08-2024
to see stock.

Description

Description

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

Certifications

CE and TUV marked. RoHS compliant.

Services

12 Months

Specifications

Specifications

200 to 900 nm
CE and TUV marked. RoHS compliant.
38.1 cm
24 sec. (96 wells), 76 sec. (384 wells)
Monochromator Bandpass
12 in.
30.5 cm
Fluorescence; Luminescence
Multi-mode Microplate Reader
0.001 OD
2
1 year
27 lb.
15 in.
Photodiode, PMT
Xenon Flash
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
≤5 nm
15 in.
Linear, Orbital, Double Orbital
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
50/60 Hz
<1% from 0 to 2.5 OD
SLXF
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
English
100/240 V
12.3 kg
38.1 cm
Full data analysis and reporting (under Gen5 control)
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers