missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CBF CO2 Incubator, with Hot Air Sterilization and Humidity Regulation

The CBF is the high-end CO2 incubator with humidity control that is suitable for all sensitive incubation applications in GMP environments. It ensures consistent results including in cell-based assays with multiwell plates.

Brand:  BINDER™ 9640-0022


12162.98 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 122.00000kg

 View more versions of this product

Product Code. 17373703

  • £17152.00 / Each
Estimated Shipment: 24-07-2024
to see stock.

Description

Description

This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data logger, measured values can be read out in open format via USB
  • Troubleshooting system with visual and acoustic alarms
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures
Specifications

Specifications

9.4 cu. ft.
10 kg
267 L
0 to 20%
Fanless interior
None
Stainless Steel
3
Yes
90 to 95%
CO2 Incubator
CBF260-230 V-O
IR sensor
745 x 885 x 1020 mm
230 V
7 A
22 lb.
CBF260-230 V-O, with O2 control 0.2-20%, Series CBF - CO2 incubators with hot air sterilization and humidity regulation, 267 L, 230 V 1 ∼ 50/60 Hz
Stainless Steel
IR Sensor
Natural
No Lighting
1
0.2 to 20%
Nema 5-20P
Ambient +6° to 60°C
TFT touch
IR sensor
620 x 575 x 750 mm
125 kg
1.5 kW
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services