missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ proFood™ 79-340 Series Green Nitrile Gloves

Reusable gloves suitable for contact with all fatty foodstuffs.

Brand:  Ansell™ 79340090

 View more versions of this product

Product Code. 11707723

  • £29.05 / 12 pairs
In Stock
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Good mechanical and chemical properties
  • Contains no natural rubber
  • Safe for both food and workers hands
  • Flexibility, even in cold condition

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Janitorial use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrile
Gloves
79-340
Green
Cotton Flocking
Mechanical and chemical
Nitrile
9
0.3 mm
Straight
12 Pairs
79-340
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers