missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ proFood™ 79-340 Series Green Nitrile Gloves

Reusable gloves suitable for contact with all fatty foodstuffs.

£7.26 - £29.05

Specifications

Back Material Nitrile
Cuff Material Nitrile
Product Type Gloves
Style 79-340
Thickness (Metric) 0.3 mm
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12687001
View Documents
Ansell™
79340100
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11787713
View Documents
Ansell™
79340070
7 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11797713
View Documents
Ansell™
79340080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11707723
View Documents
Ansell™
79340090
9
£29.05
12 pairs
In Stock
to see stock.
Add to basket
 
Description

Description

  • Good mechanical and chemical properties
  • Contains no natural rubber
  • Safe for both food and workers hands
  • Flexibility, even in cold condition

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Janitorial use
Specifications

Specifications

Nitrile
Gloves
0.3 mm
Straight
12 Pairs
79-340
Nitrile
79-340
Green
Cotton Flocking
Mechanical and chemical
Videos
SDS
Documents

Documents

Product Certifications