missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ proFood™ 79-340 Series Green Nitrile Gloves

Reusable gloves suitable for contact with all fatty foodstuffs.

Brand:  Ansell™ 79340100

 View more versions of this product

Product Code. 12687001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Good mechanical and chemical properties
  • Contains no natural rubber
  • Safe for both food and workers hands
  • Flexibility, even in cold condition

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Janitorial use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrile
Gloves
79-340
Green
Cotton Flocking
Mechanical and chemical
Nitrile
10
0.3 mm
Straight
12 Pairs
79-340
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers