Learn More
Tocris Bioscience™ ProK
Unnatural amino acid, propargyl lysine derivative
Brand:  Tocris Bioscience™ 7361/50
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18707696
  • £223.54 / 50mg
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
ProK is a lysine-based unnatural amino acid (UAA) used for genetic code expansion. It enables bioconjugation of fluorescent probes in E. coli or multicellular organisms.
Specifications
Specifications
N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride
50 mg
C10H16N2O4.HCl
1428330-91-9
Unnatural Amino Acids
0.95