missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ProK
Unnatural amino acid, propargyl lysine derivative
£65.87 - £223.54
Specifications
Chemical Name or Material | N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride |
---|---|
CAS | 1428330-91-9 |
Target | Unnatural Amino Acids |
Molecular Formula | C10H16N2O4.HCl |
Purity | 0.95 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758496
|
Tocris Bioscience™
7361/10 |
10 mg |
£65.87
10mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18707696
|
Tocris Bioscience™
7361/50 |
50 mg |
£223.54
50mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ProK is a lysine-based unnatural amino acid (UAA) used for genetic code expansion. It enables bioconjugation of fluorescent probes in E. coli or multicellular organisms.Specifications
N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride | |
Unnatural Amino Acids | |
0.95 |
1428330-91-9 | |
C10H16N2O4.HCl |