missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin Antibody (SR1366), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21891-50ul
This item is not returnable.
View return policy
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Western BlotSpecifications
Prolactin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1366 | |
Western Blot 1:500-1:2000 | |
prolactin | |
A synthesized peptide derived from human Prolactin (Uniprot #: P01236) | |
50 μg | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
5617 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |