missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin Antibody (SR1366), Novus Biologicals™
Rabbit Monoclonal Antibody
£258.00 - £340.00
Specifications
Antigen | Prolactin |
---|---|
Clone | SR1366 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18634054
|
Novus Biologicals
NBP3-21891-100ul |
100 μg |
£340.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18640815
|
Novus Biologicals
NBP3-21891-50ul |
50 μg |
£258.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Western BlotSpecifications
Prolactin | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
prolactin | |
A synthesized peptide derived from human Prolactin (Uniprot #: P01236) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1366 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5617 | |
IgG | |
Affinity purified |