missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Proof Line Weight Set

Proof Line weights are high-quality working standards for everyday use, made of high-quality materials.

Brand:  Sartorius™ YCS01-525-AC-00

 View more versions of this product

Product Code. 15499213

  • £286.00 / 1 set
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Non-magnetizable
  • Supplied with gloves, tweezers and a brush in a high-quality plastic case
  • Provided as leaf weights
  • Made of aluminum that has a density of 2,650 kg/m³
  • Without DAkkS calibration certificate
  • OIML Class M1
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Proof Line Weight Set
Aluminum
1 to 200 g
Gloves, tweezers, brush
OIML Class M1
Plastic
2,650 kg/m³
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.