missing translation for 'onlineSavingsMsg'
Learn More

Propan-1-ol, for HPLC, Fisher Chemical™

£62.15 - £125.00

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10449760
View Documents Promotion Details
Fisher Chemical
P/7486/15
1 L
£62.15
1 L
Please to purchase this item. Need a web account? Register with us today!
10111440
View Documents Promotion Details
Fisher Chemical
P/7486/17
2.5 L
£125.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO

Specifications

7
71-23-8
97°C
Liquid
MFCD00002941
2.2 mPaS at 20°C
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1g/mol
-127°C
Colorless
Amber Glass Bottle
C3H8O
25mbar at 20°C
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.10
CHEBI:28831
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Propanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications