missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Propeller Stirrer

Bochem™ Propeller Stirrer for mixing media with low and medium viscosity.

£21.10 - £22.65

Specifications

Diameter (Metric) Shaft 8 mm
View More Specs

Products 2
Product Code Brand Diameter (Metric) Fan Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Fan Length (Metric) Price Quantity & Availability  
10040811
View Documents
Bochem™
9003
50 mm 300 mm
£21.10
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
10418432
View Documents
Bochem™
9010
100 mm 400 mm
£22.65
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
Description

Description

  • Medium rpm range
  • Mixing of media with low and middle viscosity
  • Ideal for homogenising and suspensioning
  • Axial flow
Specifications

Specifications

8 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Propeller Stirrer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.