missing translation for 'onlineSavingsMsg'
Learn More
Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals Product Code.: 10397060

Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals

Product Code. 10397060
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10397060

Brand: Thermo Scientific Acros 131595000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00002354
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
Synonym propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparaseptShow MoreShow Less
PubChem CID 7175
ChEBI CHEBI:32063
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 95°C to 99°C
Color White
Flash Point 180°C
Loss on Drying 0.5% max. (5 hrs) (over silica gel)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4CO2CH2CH2CH3
Quantity 500 g
Beilstein 10, 160
Merck Index 15, 7977
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 180.2
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Propyl 4-hydroxybenzoate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465627 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number131591000,131590025,131595000,131590050
Lot NumberA0465490 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number131591000,131590025,131595000,131590050
Lot NumberA0427362 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number131591000,131590025,131595000,131590050
Lot NumberA0447041 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number131591000,131590025,131595000,131590050
Lot NumberA0460404 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number131591000,131590025,131595000,131590050
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.