missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail V, EDTA Free
Inhibits serine and cysteine proteases but not all metalloproteases.
Brand: Fisher BioReagents™ 12871640
This item is not returnable.
View return policy
Description
- Reconstitute with 1mL deionized water to make 100x concentrate
- Sufficient for 100mL of buffer
Specifications
Protease Inhibitor Cocktail | |
Lyophilized | |
1 Vial | |
AEBSF, Aprotinin, E-64 and Leupeptin |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protease Inhibitor Cocktail V (EDTA Free)
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : AEBSF HCl, Aprotinin, Leupeptin, E-64
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Protease Inhibitor Cocktail V, EDTA Free > Composition: AEBSF E-64, Aprotinin Leupeptin
Spot an opportunity for improvement?Share a Content Correction