Learn More
Thermo Scientific™ Protease Inhibitor Cocktails
Protect your sample from proteolytic degradation during cell lysis with these broad-spectrum protease inhibitor cocktails.
£259.00
Specifications
Formulation | Proprietary components in DMSO |
---|---|
Form | Liquid |
Composition | Proprietary formulation |
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
10658304
|
Thermo Scientific™
78415 |
1 mL | Protease Inhibitor Cocktail, EDTA-Free |
£259.00
1mL |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
Description
Provide broad-spectrum protection against proteolytic degradation in cell lysis procedures with Thermo Scientific™ Protease Inhibitor Cocktails.
Both ready-to-use formulations inhibit most serine, cysteine, and calpain proteases that cause general degradation of proteins in cell extracts. Metalloproteases are inhibited as well. The EDTA-free formulation is ideal for preparing samples that will be analyzed by 2D gel electrophoresis. The refrigerator-stable, 100X liquid solutions are easy to use: simply add the appropriate volume of cocktail to the sample (10μL per 1mL of cell extract) and continue with your procedure. The Protease Inhibitor can be used with animal cells and tissues, bacteria, plants and yeast.
Highlights:
- Ready-to-use, 100X liquid cocktail
- Available with EDTA (supplied in separate vial) or as an EDTA-free formulation
- Effective for suppressing proteolytic activity in detergent-based cell lysis reagents
- No loss of valuable freezer space; cocktail is stable at 4°C
Specifications
Proprietary components in DMSO | |
Liquid | |
Proprietary formulation |