missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protease Inhibitor Mix

For effective protease inhibition during protein sample prep

Brand:  Cytiva 80-6501-23

Code : GE

Additional Details : Weight : 29.48350kg


Product Code. 10625385

  • £289.00 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

Sample preparation often requires the inhibition of protease activity. This mix contains a combination of competitive and noncompetitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.

  • Inhibits >95% protease activity
  • Inhibit serine, cysteine and calpain proteases
  • No EDTA; Optionally, EDTA can be added to inhibit metalloproteases

Specifications

Specifications

Protease inhibito r (100X Solution)
Protease Inhibitor Cocktail
-20°C
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : luoride
  • UNKNOWN ACUTE-2 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 2% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers