missing translation for 'onlineSavingsMsg'
Learn More
3M™ Protective Coverall 4565 Product Code.: 17875676
Estimated Shipment: 09-06-2025
to see stock.
Add to basket

3M™ Protective Coverall 4565

Product Code. 17875676
4X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 17875676

Brand: 3M™ 7100256138

Estimated Shipment: 09-06-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays and particles.

  • Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
  • Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
  • Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
  • Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
  • Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
  • Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
  • Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens
No. of Pockets None
For Use With (Application) Controlled Environments
Cleanroom Class Class 10-100,000 (ISO 4-8)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Protective Coverall 4565 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.