missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Protein A
Protein A
Brand: Novex™ 101006
Code : 50
Additional Details : Weight : 0.02100kg
Description
Protein A purified from Staphylococcus aureus. Lyophilized, salt-free powder; no preservative added. Molecular weight is approximately 42kD.Specifications
Store in freezer -5°C to -30°C. | |
Wet Ice | |
Protein A | |
Unconjugated |
For Research Use Only. Not for use in diagnostic procedures.