missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protein A Antibody Package
For use in protein purification applications
Brand: Cytiva 29-0588-05
This item is not returnable.
View return policy
Description
Protein A package is recommended for the purification of human antibodies. Contents: HiTrap Protein A HP (1 x 1mL) and HiTrap Desalting (1 x 5mL).
Specifications
Antibody affinity chromatography | |
2°C to 8°C | |
1 Kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Desalting 1 x 5 ml; part of 'Antibody package, Protein A'
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Protein A Antibody Package > Kit contents: Protein A HP, Desalting
Spot an opportunity for improvement?Share a Content Correction