Learn More
Cytiva Protein A Sepharose™ CL-4B Medium
For immunoglobin purification and fractionation. Cytiva Protein A Sepharose™ CL-4B Medium is also well suited for immunoprecipitation.
Brand:  Cytiva 17-0963-02
 View more versions of this product
Product Code. 11554955
  • £15100.00 / 500mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Strong binding to Fc region of IgG
  • Suitable for polyclonal antibody purification
  • High capacity—20mg human IgG/mL
  • pH stability Cleaning-in-Place (CIP): 2 to 10
  • Stable in commonly used aqueous buffers, 6M guanidine-HCl, 8M urea
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20mg Human IgG/mL
4% Cross Linked Agarose
2 to 10
2°C to 8°C
Native Protein A
3mg Protein A/mL Medium
90 μm
500 mL
For immunoglobin purification and fractionation and is suitable for immunoprecipitation procedures