Learn More
Applied Biosystems™ Protein Expression Sample Prep Kit
Protein Expression Sample Prep Kit
Brand: Applied Biosystems™ 4405443
Code : 50
Additional Details : Weight : 0.05000kg
Description
The Applied Biosystems™ Protein Expression Sample Preparation Kit contains the reagents required for the preparation of target and control cell lysates for use with TaqMan™ Protein Expression Assay experiments. The Cell Lysis Reagent is a buffered detergent solution, which is combined with user-supplied protease and phosphatase inhibitors, and used to lyse adherent or nonadherent cultured cells under nondenaturing conditions. The Cell Resuspension Buffer is used to resuspend the cells before adding the lysis reagent and to dilute the lysates in preparation for the assay-binding step.Specifications
Cell Lysis Buffer | |
Contains Cell Lysis Reagent (2x) (3 x 1 mL tubes) and Cell Resuspension Buffer (1 x 25 mL bottle). Store at 2 to 8°C. | |
TaqMan™ |
For Research Use Only. Not for use in diagnostic procedures.