Learn More
Cytiva Protein G HP MultiTrap™ Plates
For small-scale purification of antibodies or enrichment of proteins of interest in prepacked 96-well filter plates
Brand: Cytiva 28-9031-35
1717.17 GBP valid until 2024-12-31
Use promo code "21963" to get your promotional price.
Code : GE
Additional Details : Weight : 0.52800kg
Includes: Four prepacked Protein G HP MultiTrap 96-well filter plates with instructions, plus a protocol, with no pretreatment of sample required, for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min.
Description
- Flexible format, with protocols for both classic elution and cross-link methods
- Formatted for elution for both electrophoresis and LC-MS analysis workflows
- Fast-binding kinetics and high capacity provide high yield
- Prepacked with Protein G Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
- Can be used with centrifugation or vacuum, manually or in automated robotic systems
- Easy scale-up with HiTrap™ Protein G HP prepacked columns
- Matrix: Highly cross-linked agarose, 6%
- Binding capacity: approx. 25mg human IgG/mL medium (0.5 to 1mg IgG/well)
- Average particle size: 34μm
- pH stability: 3 to 9 (long term); 2 to 9 (short term)
- Well volume: 800μL
- Centrifugation force: <700 x G
- Vacuum pressure: -0.5 bar (max.)
Specifications
Protein G Sepharose HP MultiTrap plate | |
4 Prepacked Protein G HP MultiTrap 96-well filter plates with instructions, plus a proto col | |
Recombinant Protein G lacking Albumin-binding region (2mg/mL medium) | |
3 to 9 (long term), 2 to 9 (short term) | |
6% Highly Cross-linked Agarose | |
Protein G Sepharose Plate |
4 Pack | |
Polypropylene, Polyethylene | |
-0.5 bar | |
34μm | |
2° to 8°C | |
Small-scale purification of Antibodies or enrichment of Proteins of interest in prepacked 96-well filter plates that are ready to use, electrophoresis or LC/MS |
Safety and Handling
- Protein G HP MultiTrap
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.