missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G Sepharose™ 4 Fast Flow Media

Product Code. p-3667126 Shop All Cytiva Products
Click to view available options
Quantity:
5 mL
25 mL
200 mL
Unit Size:
200mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 11524935

Brand: Cytiva 17061801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.

  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications

TRUSTED_SUSTAINABILITY

Specifications

Ligand Concentration 2mg Protein G/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 3 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) Used in a range of diagnostic and research applications
Ligand Recombinant Protein G lacking albumin binding region
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.