missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Gel-Loading Dye for SDS-PAGE, 2X (Dark-Purple Liq./Electrophor.), Fisher BioReagents
Brand: Fisher BioReagents BP637-5
View more versions of this product
This item is not returnable.
View return policy
Description
This dye is added to protein samples prior to electrophoresis on SDS–PAGE gels.Specifications
0.15 to 0.35 AU at 252nm, 0.25 to 0.40 AU, 0.32 to 0.47 AU | |
56-81-5, 77-86-1, 151-21-3, 7647-01-0 | |
Sodium Dodecyl Sulfate (<5%), Glycerol (<12%), Tris (<1.0%), Proprietary Component (<2%), and Water (80%). | |
Purple | |
Liquid |
Protein Gel-Loading Dye for SDS-PAGE, 2X | |
Electrophoresis | |
Poly Bottle | |
5 mL |
Safety and Handling
Product Identifier
- Protein Gel Loading Dye