missing translation for 'onlineSavingsMsg'
Learn More

Protein Gel-Loading Dye for SDS-PAGE, 2X (Dark-Purple Liq./Electrophor.), Fisher BioReagents

£120.00 - £124.00

Specifications

Absorbance 0.15 to 0.35 AU at 252nm, 0.25 to 0.40 AU, 0.32 to 0.47 AU
Chemical Name or Material Protein Gel-Loading Dye for SDS-PAGE, 2X
CAS 56-81-5, 77-86-1, 151-21-3, 7647-01-0
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Sodium Dodecyl Sulfate (<5%), Glycerol (<12%), Tris (<1.0%), Proprietary Component (<2%), and Water (80%).
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10429313
View Documents
Fisher BioReagents
BP637-1
1 mL Poly Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10376363
View Documents
Fisher BioReagents
BP637-5
5 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This dye is added to protein samples prior to electrophoresis on SDS–PAGE gels.
Specifications

Specifications

0.15 to 0.35 AU at 252nm, 0.25 to 0.40 AU, 0.32 to 0.47 AU
56-81-5, 77-86-1, 151-21-3, 7647-01-0
Sodium Dodecyl Sulfate (<5%), Glycerol (<12%), Tris (<1.0%), Proprietary Component (<2%), and Water (80%).
Liquid
Protein Gel-Loading Dye for SDS-PAGE, 2X
Electrophoresis
Purple
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

Product Identifier
  • Protein Gel Loading Dye

SDS
Documents

Documents

Product Certifications