Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
Marca: Cytiva 17-0615-01
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifica
HMW-SDS Marker Kit | |
Protein and Peptide Molecular Weight Markers | |
Designed for use with SDS electrophoresis | |
Mr 220 000 | |
Unopened vials should be stored at 4°C |
Blue | |
Solid | |
Myosin: 25μg, 220000 (Mr), rabbit muscle (source), a-2-Macroglobulin: 100μg, 170000 (Mr), bovine plasma (source), b-Galactosidase: 16μg, 116000 (Mr), E. coli (source), Transferrin | |
10 Vials |