missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
£295.00
Specifications
Product Type | Protein and Peptide Molecular Weight Markers |
---|---|
Form | Solid |
Quantity | 10 Vials |
Storage Requirements | Unopened vials should be stored at 4°C |
Product Code | Brand | Kit Contents | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Product Type | Price | Quantity & Availability | ||||
10606515
|
Cytiva
17-0446-01 |
Phosphorylase b: 67μg, 97000 (Mr), Rabbit muscle (source), Albumin: 83μg, 66000 (Mr), Bovine Serum (source), Ovalbumin: 147μg, 45000 (Mr), Chicken Egg white (source), Carbonic Anhydrase | Protein and Peptide Molecular Weight Markers |
£295.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
10764737
|
Cytiva
17-0445-01 |
Thyroglobulin: 76μg, 669000 (Mr), porcine thyroid (source), Ferritin: 50μg, 440000 (Mr), equine spleen (source), Catalase: 36μg, 232000 (Mr), bovine liver (source), Lactate dehydrogenase | Protein and Peptide Molecular Weight Markers |
£295.00
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
10136284
|
Cytiva
17-0615-01 |
Myosin: 25μg, 220000 (Mr), rabbit muscle (source), a-2-Macroglobulin: 100μg, 170000 (Mr), bovine plasma (source), b-Galactosidase: 16μg, 116000 (Mr), E. coli (source), Transferrin | Protein and Peptide Molecular Weight Markers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
Protein and Peptide Molecular Weight Markers | |
Solid | |
10 Vials | |
Unopened vials should be stored at 4°C |