Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
Brand: Cytiva 17-0446-01
259.12 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GE
Additional Details : CAS Number : 53123-88-9 Weight : 0.14580kg
Description
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
LMW-SDS Marker Kit | |
Protein and Peptide Molecular Weight Markers | |
For MW determinations by polyacrylamide gel electrophoresis (PAGE) in homogeneous or gradient gels. | |
Mr 14 000 | |
Unopened vials should be stored at 4°C |
White | |
Solid | |
Phosphorylase b: 67μg, 97000 (Mr), Rabbit muscle (source), Albumin: 83μg, 66000 (Mr), Bovine Serum (source), Ovalbumin: 147μg, 45000 (Mr), Chicken Egg white (source), Carbonic Anhydrase | |
10 Vials |