missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
£26.61 - £89.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15172069
|
Invitrogen™
00-4980-93 |
100 μL |
£26.61
100µL |
In Stock
Log in to see stock. |
|||||
15586286
|
Invitrogen™
00-4980-03 |
4 x 100 μL |
£89.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
Description
- A cocktail of Brefeldin A and Monensin
- Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
- It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
- Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
- This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
Specifications
eBioscience™ | |
Protein Transport Inhibitor Cocktail |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protein Transport Inhibitor
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.