Learn More
Novus Biologicals™ Proteinase K
Brand:  Novus Biologicals™ NB900-66727
Product Code. 18700414
  • £324.00 / 2mL
Explore more special offers
Description
Description
This is a 25X concentrated solution. For use, prepare a working solution: Place 100 uL Proteinase K solution in a glas tube, add 2.4 mL distilled water and mix. The optimal digestion conditions will vary depending on the fixation conditions and should be optimized by individual laboratory. However, incubation of tissue sections for 10-30 minutes at 37C is sufficient for in situ.
Specifications
Specifications
Sodium Azide is included at less than 0.1%. Exact amount not available.
Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)