Learn More
Thermo Scientific™ Proteinase K, recombinant, PCR grade
Degrade proteins even in the presence of detergents with Proteinase K, a broad-range endolytic protease widely used for digestion of proteins in nucleic acid preparations.
Brand: Thermo Scientific™ EO0492
101.43 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Proteinase K is an endolytic protease that cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic, or hydrophobic amino acids. The Proteinase K is classified as a serine protease. The smallest peptide to be hydrolyzed by this enzyme is a tetrapeptide.
Highlights
- Ready-to-use solution
- Active in a wide range of reaction conditions
Specifications
600 to 1000 U/mL, 14 to 22 mg/mL | |
5 x 1 mL | |
Proteinase K | |
Liquid |
Safety and Handling
- Proteinase K
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Proteinase, tritirachium album serine
For Research Use Only. Not for use in diagnostic procedures.