missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin
A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins
£517.27 - £911.42
Specifications
Product Type | Immunodepletion Resin |
---|---|
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
Includes: Immunodepletion Resin, Spin-X Filter Units
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18481280
|
R&D Systems™
IDR012-020 |
20 Tests |
£517.27
20mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18472270
|
R&D Systems™
IDR012-040 |
40 Tests |
£911.42
40mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins
Specifications
Immunodepletion Resin | |
Store the unopened product at 2 - 8° C. Do not use past expiration date. |